Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198465.3 | NP_940867.2 | 21 | 3880 | ||
XM_006724632.3 | XP_006724695.1 | 21 | 3842 | |||
XM_006724633.3 | XP_006724696.1 | 21 | 3842 | |||
XM_006724634.3 | XP_006724697.1 | 21 | 3842 | |||
XM_011530887.2 | XP_011529189.1 | 21 | 3634 | |||
GenBank mRNA | AK131549.1 | - | 21 | 4906 | ||
BX538345.1 | - | 21 | 3882 |