Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290341.2 | NP_001277270.1 | 14 | 1771 | ||
NM_001290342.2 | NP_001277271.1 | 13 | 1597 | |||
NM_001290343.1 | NP_001277272.1 | 13 | 1508 | |||
NM_178842.4 | NP_849164.2 | 13 | 1504 | |||
XM_011521355.2 | XP_011519657.1 | 14 | 1814 | |||
XM_011521357.2 | XP_011519659.1 | 14 | 1708 | |||
XM_011521358.1 | XP_011519660.1 | 14 | 1592 | |||
XM_017022002.1 | XP_016877491.1 | 13 | 1588 | |||
XM_017022003.1 | XP_016877492.1 | 13 | 1513 | |||
XM_017022004.1 | XP_016877493.1 | 12 | 1424 | |||
GenBank mRNA | BC028703.1 | - | 13 | 1585 | 4837657 | |
BC034500.1 | - | 13 | 1460 | 4838423 | ||
BC034970.1 | - | 13 | 1506 | 4822006 | ||
HY237628.1 | - | 1 | 270 |