Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308045.1 | NP_001294974.1 | 2 | 1356 | ||
NM_152699.4 | NP_689912.2 | 2 | 1356 | |||
XM_005269310.2 | XP_005269367.1 | 2 | 1356 | |||
XM_005269312.2 | XP_005269369.1 | 2 | 1356 | |||
XM_011512544.2 | XP_011510846.1 | 3 | 1401 | |||
XM_011512545.2 | XP_011510847.1 | 2 | 1356 | |||
XM_017005870.1 | XP_016861359.1 | 3 | 1356 | |||
GenBank mRNA | AF335474.1 | - | 1 | 213 | ||
AK124389.1 | - | 1 | 329 | |||
AK302305.1 | - | 2 | 1356 | |||
AK313764.1 | - | 2 | 1329 | |||
BC000958.2 | - | 1 | 453 | |||
BC008589.1 | - | 1 | 453 | |||
BC030705.2 | - | 2 | 1305 | 4824344 |