Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282745.1 | NP_001269674.1 | 3 | 221 | ||
NM_001282746.1 | NP_001269675.1 | 3 | 221 | |||
NM_173081.4 | NP_775104.2 | 3 | 221 | |||
XM_005252380.2 | XP_005252437.1 | 3 | 204 | |||
XM_011519350.2 | XP_011517652.1 | 3 | 204 | |||
XM_017015831.1 | XP_016871320.1 | 3 | 204 | |||
XM_017015832.1 | XP_016871321.1 | 3 | 204 | |||
XM_017015833.1 | XP_016871322.1 | 3 | 204 | |||
GenBank mRNA | AK057389.1 | - | 3 | 164 | ||
AK302985.1 | - | 3 | 208 | |||
AY436643.1 | - | 3 | 182 | |||
BC039312.1 | - | 3 | 214 | 5296302 | ||
DB459427.1 | - | 3 | 221 |