Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPERT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152719.2 | NP_689932.1 | 2 | 203 | ||
XM_011534971.2 | XP_011533273.1 | 2 | 1477 | |||
GenBank mRNA | AK093129.1 | - | 2 | 196 | ||
HY007057.1 | - | 2 | 195 | |||
HY017873.1 | - | 2 | 205 |