Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139178.3 | NP_631917.1 | 8 | 983 | ||
GenBank mRNA | AB042029.1 | - | 8 | 946 | ||
AK315266.1 | - | 8 | 1050 | |||
BC015155.1 | - | 3 | 269 | |||
BC067257.1 | - | 8 | 984 | 6176933 | ||
BC103812.1 | - | 8 | 721 | 40000372 | ||
BC103813.1 | - | 8 | 721 | 40000375 | ||
BC103814.2 | - | 8 | 721 | 40000376 | ||
CD364334.1 | - | 3 | 599 |