Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318539.1 | NP_001305468.1 | 2, 3 | 704 | ||
NM_152731.2 | NP_689944.2 | 2, 3 | 704 | |||
XM_011514346.2 | XP_011512648.1 | 2, 3 | 692 | |||
XM_017010406.1 | XP_016865895.1 | 2, 3 | 255 | |||
XM_017010407.1 | XP_016865896.1 | 2, 3 | 238 | |||
GenBank mRNA | AK054724.1 | - | 2, 3 | 389 | ||
AK303705.1 | - | 1, 2 | 74 | |||
BC036119.1 | - | 2, 3 | 553 | |||
DC411761.1 | - | 1, 2 | 74 |