Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173558.3 | NP_775829.2 | 4 | 645 | ||
XM_011514372.2 | XP_011512674.1 | 4 | 637 | |||
XM_011514373.2 | XP_011512675.1 | 4 | 637 | |||
XM_017010429.1 | XP_016865918.1 | 4 | 637 | |||
GenBank mRNA | AK092732.1 | - | 4 | 593 | ||
AK290419.1 | - | 5 | 638 | |||
BC023645.2 | - | 4 | 645 | 5087501 | ||
BC053655.1 | - | 5 | 590 | 5755893 |