Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286635.1 | NP_001273564.1 | 9 | 1256 | ||
NM_001286636.1 | NP_001273565.1 | 10 | 1320 | |||
NM_001286637.1 | NP_001273566.1 | 9 | 1197 | |||
NM_152734.3 | NP_689947.2 | 8 | 1156 | |||
XM_011514377.2 | XP_011512679.1 | 11 | 2019 | |||
XM_017010432.1 | XP_016865921.1 | 11 | 1473 | |||
XM_017010433.1 | XP_016865922.1 | 11 | 1474 | |||
XM_017010434.1 | XP_016865923.1 | 10 | 1712 | |||
GenBank mRNA | AJ420498.1 | - | 8 | 1162 | ||
AK058086.1 | - | 10 | 1320 | |||
AK172780.1 | - | 8 | 1065 | |||
AK300351.1 | - | 9 | 1134 | |||
AK315984.1 | - | 9 | 1106 | |||
AY358637.1 | - | 9 | 1130 | |||
BC009380.2 | - | 4 | 370 | |||
BC023605.2 | - | 8 | 1014 | |||
BC095423.1 | - | 9 | 1169 | 30348810 | ||
BC105946.1 | - | 8 | 1063 | 40032607 | ||
BC105947.1 | - | 8 | 1064 | 40032604 | ||
BC107489.1 | - | 8 | 1064 | 40032608 | ||
BX647220.1 | - | 9 | 1128 | |||
FR832871.1 | - | 9 | 1200 | |||
FR832872.1 | - | 8 | 1185 |