Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286635.1 | NP_001273564.1 | 7 | 1063 | ||
NM_001286636.1 | NP_001273565.1 | 8 | 1127 | |||
NM_001286637.1 | NP_001273566.1 | 7 | 1004 | |||
NM_152734.3 | NP_689947.2 | 6 | 963 | |||
XM_011514377.2 | XP_011512679.1 | 9 | 1826 | |||
XM_017010432.1 | XP_016865921.1 | 9 | 1280 | |||
XM_017010433.1 | XP_016865922.1 | 9 | 1281 | |||
XM_017010434.1 | XP_016865923.1 | 8 | 1519 | |||
GenBank mRNA | AJ420498.1 | - | 6 | 969 | ||
AK058086.1 | - | 8 | 1127 | |||
AK172780.1 | - | 6 | 872 | |||
AK300351.1 | - | 7 | 941 | |||
AK315984.1 | - | 7 | 913 | |||
AY358637.1 | - | 7 | 937 | |||
BC009380.2 | - | 2 | 177 | |||
BC023605.2 | - | 6 | 821 | |||
BC095423.1 | - | 7 | 976 | 30348810 | ||
BC105946.1 | - | 6 | 870 | 40032607 | ||
BC105947.1 | - | 6 | 871 | 40032604 | ||
BC107489.1 | - | 6 | 871 | 40032608 | ||
BX538108.1 | - | 6 | 856 | |||
BX647220.1 | - | 7 | 935 | |||
FR832871.1 | - | 7 | 1007 | |||
FR832872.1 | - | 6 | 992 |