Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A6E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139249.2 | NP_640342.1 | 2 | 268 | ||
GenBank mRNA | AF333758.1 | - | 2 | 319 | ||
AF354931.1 | - | 2 | 268 | |||
AF354932.1 | - | 2 | 152 | |||
AF354933.1 | - | 2 | 208 | |||
BC034588.1 | - | 2 | 182 | |||
BC069322.1 | - | 2 | 268 | 7262144 | ||
BC101783.1 | - | 2 | 328 | 8069289 |