Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286834.1 | NP_001273763.1 | 2, 3 | 350 | ||
NM_001286837.1 | NP_001273766.1 | 2, 3 | 588 | |||
NM_002936.4 | NP_002927.2 | 2, 3 | 350 | |||
GenBank mRNA | AF039652.1 | - | 2, 3 | 320 | ||
AF048994.1 | - | 2, 3 | 266 | |||
AF048995.1 | - | 2, 3 | 300 | |||
AJ224117.1 | - | 2, 3 | 239 | |||
AK075490.1 | - | 2, 3 | 299 | |||
AK096913.1 | - | 2, 3 | 506 | |||
BC002973.1 | - | 2, 3 | 309 | 3537074 | ||
BT019670.1 | - | 2, 3 | 239 | |||
CR541820.1 | - | 2, 3 | 239 | |||
DA532761.1 | - | 2, 3 | 350 |