Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WDR27 SIRNA Assays ›
Gene Symbol:
WDR27
Gene Name:
WD repeat domain 27
Gene Aliases:
-
Chromosome Location:
Chr.6: 169426420 - 169702071 on Build GRCh38
UniGene ID:
Hs.131903
Species:
Human
siRNA ID
HSS137768
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WDR27
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001202550.1
NP_001189479.1
16
2040
NM_182552.4
NP_872358.4
19
2421
XM_011535682.2
XP_011533984.1
20
2611
XM_011535684.2
XP_011533986.1
20
2611
XM_011535685.2
XP_011533987.1
20
2611
XM_011535687.2
XP_011533989.1
20
2611
XM_011535688.2
XP_011533990.1
20
2611
XM_011535689.2
XP_011533991.1
20
2611
XM_011535690.2
XP_011533992.1
20
2611
XM_011535691.2
XP_011533993.1
20
2611
XM_011535692.2
XP_011533994.1
20
2611
XM_011535693.2
XP_011533995.1
20
2611
XM_011535694.2
XP_011533996.1
20
2611
XM_017010658.1
XP_016866147.1
20
2611
XM_017010659.1
XP_016866148.1
20
2611
XM_017010660.1
XP_016866149.1
20
2611
XM_017010661.1
XP_016866150.1
20
2611
XM_017010662.1
XP_016866151.1
20
2611
XM_017010663.1
XP_016866152.1
20
2611
XM_017010664.1
XP_016866153.1
20
2611
XM_017010665.1
XP_016866154.1
20
2611
XM_017010666.1
XP_016866155.1
20
2611
XM_017010667.1
XP_016866156.1
20
2611
XM_017010668.1
XP_016866157.1
20
2611
XM_017010669.1
XP_016866158.1
14
1856
GenBank mRNA
BC131633.1
-
17
1563
40119040
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0