Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153252.4 | NP_694984.4 | 19 | 2468 | ||
XM_005262113.3 | XP_005262170.1 | 19 | 2470 | |||
XM_017029384.1 | XP_016884873.1 | 15 | 1998 | |||
XM_017029385.1 | XP_016884874.1 | 19 | 2470 | |||
GenBank mRNA | AK095887.1 | - | 2 | 341 | ||
AY497046.1 | - | 19 | 2468 | |||
AY497047.1 | - | 16 | 2400 | |||
AY497048.1 | - | 19 | 2491 | |||
AY497049.1 | - | 18 | 2461 | |||
AY497050.1 | - | 19 | 2441 | |||
AY497051.1 | - | 16 | 2028 | |||
AY497052.1 | - | 21 | 2627 | |||
AY497053.1 | - | 17 | 2216 | |||
AY497054.1 | - | 16 | 2117 | |||
AY497055.1 | - | 19 | 2402 | |||
AY497056.1 | - | 16 | 2373 | |||
AY497057.1 | - | 15 | 2272 | |||
AY497058.1 | - | 15 | 2111 | |||
AY497059.1 | - | 17 | 2482 | |||
AY497060.1 | - | 13 | 2049 | |||
BC111490.1 | - | 19 | 2418 |