Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGEF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175062.3 | NP_778232.2 | 5 | 565 | ||
GenBank mRNA | AF449764.1 | - | 4 | 753 | ||
AK093160.1 | - | 2 | 417 | |||
BC036802.1 | - | 6 | 583 | |||
BC057759.1 | - | 2 | 219 | 5268021 | ||
BF967301.1 | - | 6 | 588 |