Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153267.4 | NP_694999.3 | 13 | 2551 | ||
NR_125850.1 | 13 | 2547 | ||||
GenBank mRNA | AK304531.1 | - | 13 | 2139 | ||
BC015417.2 | - | 5 | 615 | |||
BC016383.1 | - | 13 | 2269 | 4696722 | ||
BC063634.1 | - | 13 | 2528 | 4399866 | ||
BX538309.1 | - | 2 | 3542 |