Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182758.3 | NP_877435.3 | 15 | 2803 | ||
NR_102334.1 | 15 | 3001 | ||||
XM_011521433.2 | XP_011519735.1 | 16 | 3000 | |||
XM_011521435.2 | XP_011519737.1 | 15 | 2854 | |||
XM_011521436.2 | XP_011519738.1 | 15 | 2829 | |||
XM_011521437.2 | XP_011519739.1 | 14 | 2727 | |||
XM_017022061.1 | XP_016877550.1 | 15 | 2798 | |||
GenBank mRNA | AK125130.1 | - | 1 | 414 | ||
BC101614.1 | - | 14 | 2782 | 8069120 | ||
BC101616.1 | - | 14 | 2782 | 8069122 | ||
BX537884.1 | - | 15 | 2803 | |||
BX648571.1 | - | 15 | 3003 | |||
BX648751.1 | - | 9 | 2184 |