Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185106.1 | NP_001172035.1 | 2, 3 | 1188 | ||
NM_153268.3 | NP_695000.1 | 2, 3 | 1188 | |||
GenBank mRNA | AK056141.1 | - | 2, 3 | 1188 | ||
BC121155.1 | - | 2, 3 | 942 | 40121377 | ||
BC121156.1 | - | 2, 3 | 942 | 40121378 |