Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OFCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF520800.1 | - | 5 | 382 | ||
AF520803.1 | - | 3 | 254 | |||
AF520804.1 | - | 3 | 240 | |||
AF520805.1 | - | 3 | 254 | |||
AF520806.1 | - | 3 | 262 | |||
AF520807.1 | - | 4 | 312 | |||
AF520808.1 | - | 4 | 342 | |||
AF548113.1 | - | 5 | 528 | |||
AF548114.1 | - | 113 | ||||
AF548116.1 | - | 1 | 39 | |||
AF548117.1 | - | 108 | ||||
AF548118.1 | - | 215 | ||||
AF548122.1 | - | 2 | 215 | |||
AF548123.1 | - | 2 | 215 | |||
AY309094.1 | - | 6 | 463 | |||
BC113541.1 | - | 4 | 321 | 8322593 | ||
BC113543.1 | - | 4 | 321 | 8322595 | ||
BC143837.1 | - | 5 | 333 | 9052352 |
Gene Symbol: LOC107983965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017011612.1 | XP_016867101.1 | 6 | 679 |