Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321335.1 | NP_001308264.1 | 2 | 546 | ||
NM_170746.3 | NP_734467.1 | 2 | 546 | |||
GenBank mRNA | AF536829.1 | - | 2 | 442 | ||
BC021122.1 | - | 2 | 260 | 4766360 | ||
BG714263.1 | - | 2 | 261 |