Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173039.2 | NP_766627.1 | 2 | 1017 | ||
GenBank mRNA | AB028147.1 | - | 2 | 964 | ||
AK075346.1 | - | 2 | 659 | |||
AK311096.1 | - | 3 | 414 | |||
BC040443.1 | - | 2 | 1017 | 5728157 |