Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC25A45 SIRNA Assays ›
Gene Symbol:
SLC25A45
Gene Name:
solute carrier family 25 member 45
Gene Aliases:
-
Chromosome Location:
Chr.11: 65375192 - 65383701 on Build GRCh38
UniGene ID:
Hs.661604
Species:
Human
siRNA ID
HSS138234
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC25A45
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077241.2
NP_001070709.2
5
523
NM_001278250.2
NP_001265179.2
4
734
NM_001278251.2
NP_001265180.2
9
1380
NM_001300820.1
NP_001287749.1
4
620
NM_182556.3
NP_872362.3
6
578
XM_006718509.3
XP_006718572.1
5
1188
XM_006718510.3
XP_006718573.1
7
716
XM_011544943.2
XP_011543245.1
6
1287
XM_011544944.2
XP_011543246.1
5
617
XM_011544947.2
XP_011543249.1
5
806
XM_011544949.2
XP_011543251.1
7
635
XM_017017562.1
XP_016873051.1
5
806
XM_017017563.1
XP_016873052.1
6
2053
XM_017017564.1
XP_016873053.1
6
2052
XM_017017565.1
XP_016873054.1
6
1317
XM_017017566.1
XP_016873055.1
6
672
XM_017017567.1
XP_016873056.1
7
678
XM_017017568.1
XP_016873057.1
6
501
XM_017017569.1
XP_016873058.1
8
1380
XM_017017570.1
XP_016873059.1
4
1003
XM_017017571.1
XP_016873060.1
4
373
GenBank mRNA
AK090434.1
-
1
2635
AK091190.1
-
9
1380
AK124022.1
-
1
981
AY597807.1
-
4
185
BC036869.1
-
5
486
5228549
BX648573.1
-
2
5408
Back To Top
More Information
Panther Classification:
Molecular Function -
transporter
Gene Ontology Categories:
Function(s)
Process(es)
translation
transmembrane transport
structural constituent of ribosome
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0