Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318812.1 | NP_001305741.1 | 4 | 683 | ||
NM_001318813.1 | NP_001305742.1 | 4 | 785 | |||
NM_001318814.1 | NP_001305743.1 | 5 | 867 | |||
NM_001318815.1 | NP_001305744.1 | 4 | 628 | |||
NM_001318816.1 | NP_001305745.1 | 4 | 953 | |||
NM_173810.3 | NP_776171.1 | 3 | 871 | |||
GenBank mRNA | AF451992.1 | - | 3 | 812 | ||
AF451993.1 | - | 3 | 812 | |||
BC032123.2 | - | 3 | 871 | 4853202 | ||
BC053665.1 | - | 4 | 623 | 6153096 | ||
BQ050575.1 | - | 4 | 750 | |||
CN430374.1 | - | 4 | 566 |