Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173598.4 | NP_775869.3 | 16 | 2382 | ||
XM_011538224.2 | XP_011536526.1 | 16 | 2711 | |||
XM_011538225.2 | XP_011536527.1 | 16 | 3176 | |||
XM_011538228.2 | XP_011536530.1 | 14 | 1541 | |||
XM_017019208.1 | XP_016874697.1 | 16 | 2717 | |||
XM_017019210.1 | XP_016874699.1 | 11 | 1167 | |||
GenBank mRNA | AY345972.1 | - | 14 | 2051 | ||
BC107106.1 | - | 14 | 2046 | |||
BC107107.1 | - | 14 | 2046 | |||
BC127603.1 | - | 14 | 2051 | 40019272 |