Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130918.1 | NP_001124390.1 | 14 | 2200 | ||
NM_173623.3 | NP_775894.2 | 7 | 1288 | |||
XM_017024491.1 | XP_016879980.1 | 14 | 2343 | |||
XM_017024492.1 | XP_016879981.1 | 13 | 2456 | |||
XM_017024493.1 | XP_016879982.1 | 15 | 2410 | |||
XM_017024494.1 | XP_016879983.1 | 14 | 2345 | |||
XM_017024495.1 | XP_016879984.1 | 13 | 3073 | |||
XM_017024496.1 | XP_016879985.1 | 9 | 1483 | |||
GenBank mRNA | AK093127.1 | - | 6 | 1152 | ||
AK097392.1 | - | 14 | 2112 | |||
AK302952.1 | - | 14 | 2146 | |||
BC033563.1 | - | 7 | 1289 | 4825815 | ||
BC041368.2 | - | 14 | 3196 |