Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083961.1 | NP_001077430.1 | 19 | 2347 | ||
NM_173636.4 | NP_775907.4 | 19 | 2347 | |||
XM_005258809.2 | XP_005258866.1 | 19 | 2347 | |||
XM_011526837.1 | XP_011525139.1 | 19 | 2332 | |||
XM_011526838.1 | XP_011525140.1 | 19 | 2347 | |||
XM_011526839.1 | XP_011525141.1 | 17 | 1996 | |||
XM_011526840.2 | XP_011525142.1 | 12 | 1342 | |||
XM_011526841.2 | XP_011525143.1 | 10 | 963 | |||
XM_011526842.1 | XP_011525144.1 | 9 | 795 | |||
XM_011526843.1 | XP_011525145.1 | 2 | 122 | |||
XM_011526844.2 | XP_011525146.1 | 2 | 119 | |||
XM_017026665.1 | XP_016882154.1 | 19 | 2347 | |||
GenBank mRNA | BC017261.2 | - | 1 | 56 | ||
BC058939.1 | - | 19 | 2291 | |||
BQ880028.1 | - | 6 | 516 | |||
BX647726.1 | - | 20 | 2408 |