Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207348.2 | NP_997231.1 | 1 | 424 | ||
XM_011541292.2 | XP_011539594.1 | 1 | 1650 | |||
XM_011541293.1 | XP_011539595.1 | 1 | 1650 | |||
XM_017001083.1 | XP_016856572.1 | 1 | 1650 | |||
GenBank mRNA | BC027998.1 | - | 1 | 424 | 5189944 |