Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173825.3 | NP_776186.2 | 1 | 51 | ||
XM_006713595.2 | XP_006713658.1 | 1 | 35 | |||
XM_017006227.1 | XP_016861716.1 | 1 | 35 | |||
XM_017006228.1 | XP_016861717.1 | 1 | 35 | |||
GenBank mRNA | AK293085.1 | - | 1 | 35 | ||
BC020832.1 | - | 1 | 30 | 4771781 | ||
BX647559.1 | - | 1 | 48 | |||
BX647593.1 | - | 1 | 51 | |||
BX647664.1 | - | 1 | 35 |