Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207352.3 | NP_997235.3 | 2 | 602 | ||
XM_005262935.3 | XP_005262992.1 | 2 | 387 | |||
XM_017008037.1 | XP_016863526.1 | 2 | 446 | |||
GenBank mRNA | AK122600.1 | - | 2 | 337 | ||
AK126473.1 | - | 3 | 271 | |||
AY422002.1 | - | 2 | 602 | |||
BC060857.1 | - | 2 | 468 | 30333559 |