Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173667.3 | NP_775938.1 | 3 | 163 | ||
XM_017009378.1 | XP_016864867.1 | 1 | 2582 | |||
XM_017009379.1 | XP_016864868.1 | 1 | 2582 | |||
XM_017009380.1 | XP_016864869.1 | 1 | 2582 | |||
XM_017009381.1 | XP_016864870.1 | 1 | 2582 | |||
XM_017009382.1 | XP_016864871.1 | 2 | 84 | |||
GenBank mRNA | AK094862.1 | - | 3 | 135 | ||
BC111982.1 | - | 2 | 89 | 8327450 | ||
BC112008.1 | - | 2 | 89 | 8327476 |