Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173834.3 | NP_776195.2 | 2, 3 | 471 | ||
XM_011530923.2 | XP_011529225.1 | 2, 3 | 879 | |||
XM_011530924.1 | XP_011529226.1 | 2, 3 | 253 | |||
XM_017029450.1 | XP_016884939.1 | 2, 3 | 253 | |||
GenBank mRNA | AK095555.1 | - | 2, 3 | 253 | ||
AK292835.1 | - | 2, 3 | 213 | |||
BC012469.1 | - | 2, 3 | 209 | 4452699 |