Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176891.4 | NP_795372.1 | 1 | 1034 | ||
GenBank mRNA | AY190045.1 | - | 1 | 421 | ||
AY358570.1 | - | 1 | 1035 | |||
BC100871.2 | - | 1 | 421 | 40003063 | ||
BC100872.2 | - | 1 | 421 | 40003065 | ||
BC100873.1 | - | 1 | 421 | 40003066 |