Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD13D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207354.2 | NP_997237.2 | 8 | 1024 | ||
NR_030767.1 | 9 | 1107 | ||||
XM_011544985.1 | XP_011543287.1 | 8 | 967 | |||
XM_011544986.1 | XP_011543288.1 | 9 | 1107 | |||
XM_011544987.2 | XP_011543289.1 | 8 | 941 | |||
XM_011544988.2 | XP_011543290.1 | 3 | 287 | |||
GenBank mRNA | AK027313.1 | - | 1 | 955 | ||
AK057566.1 | - | 1 | 2481 | |||
AK057681.1 | - | 1 | 1638 | |||
AK096277.1 | - | 8 | 937 | |||
AK124721.1 | - | 9 | 1104 | |||
AK126438.1 | - | 7 | 1021 | |||
BC110419.1 | - | 8 | 997 | 5210878 | ||
BC121024.1 | - | 8 | 898 | 40117420 | ||
BC121025.2 | - | 8 | 898 | 40117423 |