Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163857.1 | NP_001157329.1 | 4 | 755 | ||
NM_178552.3 | NP_848647.1 | 4 | 736 | |||
XM_011530165.2 | XP_011528467.1 | 5 | 968 | |||
XM_011530166.1 | XP_011528468.1 | 4 | 711 | |||
GenBank mRNA | BC042635.1 | - | 4 | 698 | 5171722 | |
BE502000.1 | - | 3 | 293 | |||
CR456416.1 | - | 4 | 721 |