Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135197.1 | NP_001128669.1 | 8 | 1004 | ||
NM_178173.3 | NP_835467.2 | 10 | 1302 | |||
XM_006713124.3 | XP_006713187.1 | 8 | 1179 | |||
XM_011533670.2 | XP_011531972.1 | 7 | 1013 | |||
XM_011533671.2 | XP_011531973.1 | 7 | 1007 | |||
XM_011533672.2 | XP_011531974.1 | 8 | 1372 | |||
XM_011533673.2 | XP_011531975.1 | 8 | 1404 | |||
GenBank mRNA | AK058049.1 | - | 10 | 1193 | ||
AK303033.1 | - | 8 | 1004 | |||
BC036202.1 | - | 10 | 1302 | 4836976 |