Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178564.3 | NP_848659.2 | 15 | 1420 | ||
XM_017013378.1 | XP_016868867.1 | 15 | 1420 | |||
XM_017013379.1 | XP_016868868.1 | 15 | 1420 | |||
XM_017013380.1 | XP_016868869.1 | 15 | 1420 | |||
XM_017013381.1 | XP_016868870.1 | 15 | 1420 | |||
XM_017013382.1 | XP_016868871.1 | 15 | 1420 | |||
XM_017013383.1 | XP_016868872.1 | 15 | 1420 | |||
GenBank mRNA | AF318376.1 | - | 5 | 760 | ||
AK094017.1 | - | 2 | 129 | |||
AK293943.1 | - | 15 | 1420 | |||
AK308967.1 | - | 2 | 111 | |||
AL137662.1 | - | 13 | 1349 | |||
AL834530.1 | - | 8 | 659 | |||
AY277601.1 | - | 5 | 747 | |||
BC071605.1 | - | 14 | 1583 | |||
BC113873.1 | - | 12 | 1424 | 40053695 | ||
BC114481.1 | - | 13 | 1321 | |||
CR457350.1 | - | 7 | 551 |