Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272046.1 | NP_001258975.1 | 4 | 481 | ||
NM_001320804.1 | NP_001307733.1 | 4 | 423 | |||
XM_011539754.2 | XP_011538056.1 | 4 | 480 | |||
XM_011539757.2 | XP_011538059.1 | 4 | 753 | |||
XM_017016177.1 | XP_016871666.1 | 4 | 233 | |||
XM_017016179.1 | XP_016871668.1 | 4 | 186 | |||
XM_017016181.1 | XP_016871670.1 | 4 | 230 | |||
GenBank mRNA | AJ536328.2 | - | 4 | 405 | ||
AJ616914.1 | - | 4 | 405 | |||
AK122716.1 | - | 4 | 476 | |||
AK127756.1 | - | 4 | 312 | |||
AY572972.1 | - | 4 | 243 | |||
BC143380.1 | - | 4 | 183 | |||
DA170480.1 | - | 4 | 481 |