Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181536.1 | NP_853514.1 | 7, 8 | 1148 | ||
XM_017023201.1 | XP_016878690.1 | 7, 8 | 1571 | |||
XM_017023202.1 | XP_016878691.1 | 7, 8 | 1571 | |||
XM_017023203.1 | XP_016878692.1 | 7, 8 | 1571 | |||
XM_017023204.1 | XP_016878693.1 | 7, 8 | 1571 | |||
GenBank mRNA | AY164485.1 | - | 7, 8 | 1148 |