Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1211L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207362.2 | NP_997245.2 | 6 | 954 | ||
XM_005263923.3 | XP_005263980.1 | 6 | 934 | |||
XM_005263924.3 | XP_005263981.1 | 6 | 934 | |||
XM_005263926.3 | XP_005263983.1 | 6 | 819 | |||
XM_011511093.1 | XP_011509395.1 | 8 | 1093 | |||
XM_011511094.1 | XP_011509396.1 | 7 | 991 | |||
XM_011511095.2 | XP_011509397.1 | 6 | 761 | |||
XM_011511096.1 | XP_011509398.1 | 3 | 371 | |||
XM_017003971.1 | XP_016859460.1 | 6 | 937 | |||
XM_017003972.1 | XP_016859461.1 | 7 | 1041 | |||
XM_017003973.1 | XP_016859462.1 | 6 | 947 |