Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207366.2 | NP_997249.2 | 8 | 979 | ||
XM_011515373.2 | XP_011513675.1 | 8 | 963 | |||
GenBank mRNA | AI337138.1 | - | 1 | 15 | ||
AK126048.1 | - | 3 | 267 | |||
AK301928.1 | - | 8 | 942 | |||
DB087561.1 | - | 3 | 267 |