Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NMNAT3 SIRNA Assays ›
Gene Symbol:
NMNAT3
Gene Name:
nicotinamide nucleotide adenylyltransferase 3
Gene Aliases:
FKSG76, PNAT-3, PNAT3
Chromosome Location:
Chr.3: 139560180 - 139678050 on Build GRCh38
UniGene ID:
Hs.208673
Species:
Human
siRNA ID
HSS139489
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NMNAT3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320510.1
NP_001307439.1
5
891
NM_001320511.1
NP_001307440.1
5
643
NM_001320512.1
NP_001307441.1
4
609
NM_001320513.1
NP_001307442.1
4
609
NM_178177.4
NP_835471.1
3
460
XM_011512790.1
XP_011511092.1
2
35
XM_011512791.2
XP_011511093.1
2
334
XM_017006328.1
XP_016861817.1
6
908
XM_017006329.1
XP_016861818.1
6
905
XM_017006330.1
XP_016861819.1
5
868
XM_017006331.1
XP_016861820.1
6
981
XM_017006332.1
XP_016861821.1
6
4922
XM_017006333.1
XP_016861822.1
4
771
XM_017006334.1
XP_016861823.1
4
768
XM_017006335.1
XP_016861824.1
5
620
XM_017006336.1
XP_016861825.1
4
580
XM_017006337.1
XP_016861826.1
3
483
XM_017006338.1
XP_016861827.1
2
35
XM_017006339.1
XP_016861828.1
5
806
XM_017006340.1
XP_016861829.1
4
775
XM_017006341.1
XP_016861830.1
4
524
XM_017006342.1
XP_016861831.1
5
630
XM_017006343.1
XP_016861832.1
4
590
XM_017006344.1
XP_016861833.1
3
490
GenBank mRNA
AF345564.1
-
2
159
BC034374.1
-
3
408
5185621
BC036218.1
-
4
576
5270916
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotidyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
NAD biosynthetic process
response to wounding
NAD metabolic process
'de novo' NAD biosynthetic process from aspartate
nicotinamide-nucleotide adenylyltransferase activity
nicotinate-nucleotide adenylyltransferase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-jgtjs:80/10.239.162.121:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0