Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143679.1 | NP_001137151.1 | 1, 2 | 163 | ||
NM_001143680.1 | NP_001137152.1 | 1, 2 | 163 | |||
NM_001143681.1 | NP_001137153.1 | 1, 2 | 163 | |||
NM_015917.2 | NP_057001.1 | 1, 2 | 163 | |||
GenBank mRNA | AF068287.1 | - | 1, 2 | 150 | ||
AF070657.1 | - | 1, 2 | 121 | |||
AF087849.1 | - | 1, 2 | 135 | |||
AK000640.1 | - | 1, 2 | 121 | |||
AK095263.1 | - | 1, 2 | 105 | |||
AK289570.1 | - | 1, 2 | 96 | |||
AK289580.1 | - | 1, 2 | 101 | |||
AK295592.1 | - | 1, 2 | 151 | |||
AK299968.1 | - | 1, 2 | 81 | |||
AL136938.1 | - | 1, 2 | 149 | |||
AY520571.1 | - | 1, 2 | 132 | |||
BC001231.1 | - | 1, 2 | 110 | 3460150 | ||
BC050715.1 | - | 1, 2 | 83 | 6061265 | ||
BC063425.1 | - | 1, 2 | 92 | 5218251 | ||
BQ436828.1 | - | 1, 2 | 163 | |||
CR533445.1 | - | 1, 2 | 78 |