Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153614.3 | NP_705842.2 | 6 | 1431 | ||
XM_005273984.3 | XP_005274041.1 | 5 | 796 | |||
XM_011545004.2 | XP_011543306.1 | 8 | 1682 | |||
XM_011545005.2 | XP_011543307.1 | 7 | 1522 | |||
XM_011545007.2 | XP_011543309.1 | 6 | 1207 | |||
XM_011545009.2 | XP_011543311.1 | 8 | 1356 | |||
XM_011545015.2 | XP_011543317.1 | 3 | 549 | |||
XM_017017675.1 | XP_016873164.1 | 7 | 952 | |||
XM_017017676.1 | XP_016873165.1 | 8 | 1532 | |||
XM_017017677.1 | XP_016873166.1 | 7 | 1401 | |||
XM_017017678.1 | XP_016873167.1 | 6 | 1212 | |||
GenBank mRNA | AF516185.1 | - | 3 | 1403 | ||
AK058095.1 | - | 3 | 1403 | |||
AK128589.1 | - | 8 | 3476 | |||
AY138810.2 | - | 6 | 1431 | |||
AY325766.1 | - | 6 | 724 | |||
BC127689.1 | - | 1 | 538 |