Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175078.2 | NP_778253.2 | 2 | 716 | ||
XM_011538288.2 | XP_011536590.1 | 2 | 478 | |||
XM_011538289.2 | XP_011536591.1 | 2 | 777 | |||
GenBank mRNA | AJ564104.1 | - | 2 | 716 | ||
BC118598.1 | - | 3 | 801 | 40108170 | ||
BC122558.1 | - | 2 | 716 | 40108176 |