Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MROH7-TTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037639.2 | 3 | 1150 | |||
NR_037640.2 | 3 | 1045 | ||||
NR_037641.2 | 3 | 1045 | ||||
GenBank mRNA | BX648088.1 | - | 3 | 983 | ||
CR749830.1 | - | 3 | 1102 |
Gene Symbol: MROH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039464.3 | NP_001034553.2 | 3 | 1045 | ||
NR_026782.2 | 3 | 1039 | ||||
NR_111931.1 | 3 | 1045 | ||||
GenBank mRNA | AK093272.1 | - | 3 | 984 | ||
AK130047.1 | - | 2 | 1534 | |||
AK302446.1 | - | 3 | 992 | |||
AL832492.1 | - | 1 | 272 | |||
BC095478.1 | - | 2 | 827 | |||
BC126126.1 | - | 1 | 745 | 8991842 | ||
BC143891.1 | - | 1 | 745 | 9052408 | ||
BC144516.1 | - | 3 | 983 |