Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130711.1 | NP_001124183.1 | 3 | 329 | ||
NM_207375.2 | NP_997258.1 | 3 | 329 | |||
XM_011520654.2 | XP_011518956.1 | 3 | 378 | |||
XM_011520656.2 | XP_011518958.1 | 3 | 378 | |||
XM_011520657.2 | XP_011518959.1 | 3 | 377 | |||
GenBank mRNA | AY359126.1 | - | 3 | 295 | ||
EF127467.1 | - | 3 | 329 | |||
EU095393.1 | - | 3 | 256 | |||
EU095394.1 | - | 3 | 256 |