Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145199.1 | NP_001138671.1 | 6 | 620 | ||
GenBank mRNA | AB735446.1 | - | 6 | 560 | ||
AB735447.1 | - | 5 | 584 | |||
AJ311377.1 | - | 6 | 486 | |||
AJ311378.1 | - | 6 | 394 | |||
AK056999.1 | - | 6 | 492 | |||
BC013920.1 | - | 6 | 622 | 3838595 | ||
BC045812.1 | - | 6 | 584 | 4829271 | ||
BC061920.1 | - | 6 | 546 | 4413691 | ||
BC067897.1 | - | 6 | 594 | 5295847 | ||
BC126980.1 | - | 5 | 314 | 40108673 | ||
BC127012.1 | - | 5 | 314 | 40108672 | ||
BC131552.1 | - | 5 | 314 |