Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203347.1 | NP_976222.1 | 5 | 480 | ||
XM_011518672.1 | XP_011516974.1 | 5 | 408 | |||
GenBank mRNA | AY358618.1 | - | 5 | 480 | ||
BC093893.1 | - | 5 | 475 | 7939738 | ||
BC093895.1 | - | 5 | 475 | 7939740 |