Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203303.2 | NP_976048.1 | 1 | 193 | ||
GenBank mRNA | BC021176.2 | - | 1 | 193 | 5269882 |