Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212559.2 | NP_997724.2 | 1 | 879 | ||
XM_005262130.2 | XP_005262187.1 | 1 | 1467 | |||
XM_011530954.2 | XP_011529256.1 | 1 | 1467 | |||
GenBank mRNA | AK296711.1 | - | 1 | 1403 | ||
AY534239.1 | - | 1 | 451 | |||
AY589511.1 | - | 1 | 879 | |||
BC137010.1 | - | 1 | 521 | 9021003 | ||
BC137011.1 | - | 1 | 521 | 9021004 |